Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Individual Analytical Precision Weights, Class 1

Catalog No. 023191P Shop All Troemner Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
300 mg
500 mg
1 g
3 g
5 g
10 g
50 g
100 g
200 g
500 g
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass
02-319-1P 10 mg
02-319-1J 200 mg
02-319-1H 500 mg
02-319-1K 100 mg
02-319-1B 50 g
02-319-1E 5 g
02-319-1T 1 mg
02-319-1Q 5 mg
02-319-1D 10 g
02-319-1L 50 mg
02-215-77 200 g
02-214-021 300 mg
02-215-95 500 g
02-319-1R 2 mg
02-319-1A 100 g
02-319-1G 1 g
02-214-022 3 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 02-319-1P Supplier Troemner™ Supplier No. 80781109
Add to Cart
Add to Cart

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Can be used as a reference standard in calibrating other weights
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.1 mg to 0.01mg

Certifications

A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements

TRUSTED_SUSTAINABILITY

Specifications

Product Type One Piece Metric Analytical Sheet Weight
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 10 mg
Density 7.95g/cm3 at 20°C
Mass 10 mg
Shape Flat with one end turned up for easy handling
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.