Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Individual Analytical Precision Weights, Class 2 with Traceable Certificate

Catalog No. 02214083 Shop All Troemner Products
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
50 mg
200 mg
300 mg
500 mg
1 g
3 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass
02-214-083 5 kg
02-217-2FT 50 g
02-217-4AT 1 mg
02-217-4BT 2 mg
02-214-080 500 g
02-217-2GT 100 g
02-217-2DT 10 g
02-217-2AT 1 g
02-214-079 3 g
02-214-078 300 mg
02-214-082 2 kg
02-217-2CT 5 g
02-214-081 1 kg
02-217-3FT 500 mg
02-217-3CT 50 mg
02-225-550 200 g
02-217-2ET 20 g
02-217-4CT 5 mg
02-217-3AT 10 mg
02-225-513 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 02-214-083 Supplier Troemner™ Supplier No. 80858085
Add to Cart
Add to Cart

Designed for your most demanding weighing applications

Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.

  • Precision Analytical Individual Weights are made from the finest materials and are manufactured in sizes ranging from 1mg through 50kg
  • Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
  • Fractional weights 3mg and below are made of aluminum
  • Weights supplied in a rugged, precision-fit, high-quality polycarbonate case
  • Can be also be used as reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Used to calibrate weighing instruments for important commercial transactions like gold and precious stones
  • Calibrate Class 3 weights
TRUSTED_SUSTAINABILITY

Specifications

Product Type Two-Piece Metric Analytical Weight
Certifications/Compliance Traceable certificate, ISO9001
Classification ASTM Class 2
Material Stainless Steel
Weight (Metric) 5 kg
Density 7.95g/cm3 at 20°C
Mass 5 kg
Shape Cylindrical with Handling Knob
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.