Learn More
Troemner™ Individual Analytical Precision Weights, Class 2 with Traceable Certificate
Shop All Troemner ProductsDescription
Weights are designed so the specifications on surface finish, markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances decrease in order to enable the weight to maintain its tolerance over time.
- Precision Analytical Individual Weights are made from the finest materials and are manufactured in sizes ranging from 1mg through 50kg
- Fractional weights 5mg to 500mg are made of stainless steel (30mg weights are aluminum)
- Fractional weights 3mg and below are made of aluminum
- Weights supplied in a rugged, precision-fit, high-quality polycarbonate case
- Can be also be used as reference standards as well as calibrating Class I balances as specified in OIML R 76
- Used to calibrate weighing instruments for important commercial transactions like gold and precious stones
- Calibrate Class 3 weights
Specifications
Specifications
| Product Type | One Piece Metric Analytical Sheet Weight |
| Certifications/Compliance | Traceable certificate, ISO9001 |
| Classification | ASTM Class 2 |
| Material | Stainless Steel |
| Weight (Metric) | 500 mg |
| Density | 7.95g/cm3 at 20°C |
| Mass | 500 mg |
| Shape | Flat with one end turned up for easy handling |
| Includes | Polycarbonate case |
| Quantity | Each |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.