Learn More
Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate
Shop All Troemner ProductsDescription
Designed for your most demanding weighing applications, Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
- Weights 1mg through 2kg are supplied in a high quality polycarbonate case
- Reference standards as well as calibrating Class I balances as specified in OIML R 76
- Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
Order Info
Protective cases that are supplied with weights can be ordered separately.
Specifications
Specifications
| Product Type | Metric Analytical Weight |
| Certifications/Compliance | Traceable certificate, ISO9001 |
| Classification | ASTM Class 3 |
| Material | Stainless Steel |
| Weight (Metric) | 1 kg |
| Density | 7.95g/cm3 at 20°C |
| Mass | 1 kg |
| Shape | Cylindrical with Handling Knob |
| Includes | Polycarbonate case |
| Quantity | Each |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.