Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Individual Analytical Precision Weights, Class 3 with Traceable Certificate

Catalog No. 02225526 Shop All Troemner Products
Change view
Click to view available options
Mass:
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
300 mg
2 g
3 g
200 g
1 kg
2 kg
5 kg
Material:
Aluminum
Stainless Steel
Shape:
Cylindrical with Handling Knob
Flat with one side turned up
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass Material Shape
02-225-526 5 mg Stainless Steel Flat with one side turned up
02-225-518 50 mg Stainless Steel Flat with one side turned up
02-225-529 2 mg Aluminum Flat with one side turned up
02-215-30DT 100 mg Stainless Steel Flat with one side turned up
02-214-084 300 mg Stainless Steel Flat with one side turned up
02-225-509 2 g Stainless Steel Cylindrical with Handling Knob
02-214-086 3 g Stainless Steel Cylindrical with Handling Knob
02-214-090 5 kg Stainless Steel Cylindrical with Handling Knob
02-225-514 200 mg Stainless Steel Flat with one side turned up
02-214-088 1 kg Stainless Steel Cylindrical with Handling Knob
02-214-089 2 kg Stainless Steel Cylindrical with Handling Knob
02-225-523 10 mg Stainless Steel Flat with one side turned up
02-214-087 200 g Stainless Steel Cylindrical with Handling Knob
02-225-521 20 mg Stainless Steel Flat with one side turned up
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 02-225-526 Supplier Troemner™ Supplier No. 80858617
Add to Cart
Add to Cart

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Designed for your most demanding weighing applications, Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01g to 0.001g
  • Weights 1mg through 2kg are supplied in a high quality polycarbonate case
  • Reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g

Order Info

Protective cases that are supplied with weights can be ordered separately.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Metric Analytical Weight
Certifications/Compliance Traceable certificate, ISO9001
Classification ASTM Class 3
Material Stainless Steel
Weight (Metric) 5 mg
Density 7.95g/cm3 at 20°C
Mass 5 mg
Shape Flat with one side turned up
Includes Polycarbonate case
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.