Learn More
Description
Includes
Comes with living material.
Easy to use. Easy to raise.
- S28869 comes with coupon, pop-up habitat, feeder and instructions
- S28870 comes with live butterfly larvae, pop-up habitat, feeder and instructions
Perfect for all ages.
Specifications
Specifications
| For Use With (Application) | Allow students to work hands on with living butterflies. |
| Quantity | Ea. |
| Includes | Live butterfly larvae and complete instructions |
| Sufficient For | Class of 30 students |
| Product Type | Individual Butterfly Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

