Learn More
Research Products International Corp Individual Feeder Dish & Screen (Mouse)
SDP
Catalog No. 501978046
Encompass
The Individual Feeder Dish and Screen are designed to provide precise control of mouse feeding in diet and nutritional studies. Constructed from high-quality stainless steel, these components are durable, corrosion-resistant, and engineered for repeated laboratory use. The smooth surfaces ensure quick, thorough cleaning and are fully autoclavable for reliable sterilization. By limiting feed access, the dish and screen enable accurate measurement of intake while maintaining hygienic conditions in the enclosure.
Catalog No. 50-197-8046 Supplier Research Products International Corp Supplier No. 929120
The Individual Feeder Dish and Screen are designed to provide precise control of mouse feeding in diet and nutritional studies. Constructed from high-quality stainless steel, these components are durable, corrosion-resistant, and engineered for repeated laboratory use. The smooth surfaces ensure quick, thorough cleaning and are fully autoclavable for reliable sterilization. By limiting feed access, the dish and screen enable accurate measurement of intake while maintaining hygienic conditions in the enclosure.