Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Individual Instruction: Plant Tissue Macronutrients
SCI_ED

Catalog No. S02003 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S02003 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02003 Supplier Innovating Science™ Supplier No. IS1520
Add to Cart
Add to Cart

Plants need a variety of nutrients and minerals to grow and maintain overall health. Test for the three main nutrients that plants require to survive and thrive: nitrogen, phosphorus, and potassium.

Plants need a variety of nutrients and minerals to grow and maintain overall health. The three main nutrients that plants require to survive and thrive are nitrogen, phosphorus, and potassium (also known as NPK). In this activity, students will extract nutrients from plants of your choice and perform analyses to determine if nitrogen, phosphorus, and potassium concentration is abundant, adequate, or deficient.

Specifications

Grade 6+
Class Size 1 Lab Group
Product Type Macronutrient Plant Tissue Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.