Learn More
Research Products International Corp Individual Poly Enclosure Set, Rat
SDP
Catalog No. 501977828
Encompass
This durable polycarbonate enclosure is designed to comfortably house a rat while enabling controlled feeding for diet and nutritional studies. Used in combination with Dish and Screen Set (SKU 929360).  It provides a dependable system for precise feed regulation in animal model research. The enclosure is lightweight yet impact-resistant, cleans quickly, and is fully autoclavable to ensure sterility and long-term laboratory use.
Catalog No. 50-197-7828 Supplier Research Products International Corp Supplier No. 929110
This durable polycarbonate enclosure is designed to comfortably house a rat while enabling controlled feeding for diet and nutritional studies. Used in combination with Dish and Screen Set (SKU 929360).  It provides a dependable system for precise feed regulation in animal model research. The enclosure is lightweight yet impact-resistant, cleans quickly, and is fully autoclavable to ensure sterility and long-term laboratory use.