Learn More
MSC Value Collection Round Steel Spacer Block, 0.17 in., Grade B, 1 ea.
Catalog No. 5000516912
Encompass
This round steel gage block is designed for precise measurement and calibration applications. Featuring a Grade B accuracy rating, this spacer block is constructed from durable steel to ensure reliable performance in layout and machine setup tasks.
  • Construction from high-quality steel
  • Grade B accuracy for reliable measurements
  • Round shape design
  • Nominal size of 0.17 in.
Catalog No. 50-005-16912 Supplier MSC Supplier No. 06464333
This round steel gage block is designed for precise measurement and calibration applications. Featuring a Grade B accuracy rating, this spacer block is constructed from durable steel to ensure reliable performance in layout and machine setup tasks.
  • Construction from high-quality steel
  • Grade B accuracy for reliable measurements
  • Round shape design
  • Nominal size of 0.17 in.