Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRANDTECH™ BRAND™ Individual Stand for Transferpette™ Electronic Pipets

Catalog No. 13689217 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Transferpette Electronic Single Channel Pipette (500 to 5000 μL)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-689-217 Transferpette Electronic Single Channel Pipette (500 to 5000 μL)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-689-217 Supplier BRANDTECH™ BRAND™ Supplier No. 705386
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Also suitable for corresponding models of Transferpette™ and Transferpette S pipets.

  • Available for two different capacities
  • Provided individually
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Color Transparent/White
Disposable Reusable
For Use With Transferpette/Transferpette S
Material Polypropylene, Stainless Steel
Number of Channels 1
Sterility Non-sterile
Type Pipette Stand
For Use With (Equipment) Transferpette Electronic Single Channel Pipette (500 to 5000 μL)
Width (English) 4.5 in.
Height (English) 3.12 in.
Height (Metric) 7.92 cm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.