Learn More
GENEPATH DIAGNOSTICS INC Individual VanA,VanB,VanC1,VanC2/3 quantitative standards for assay calibration and validation. Provided at high (10,000 cps/µL), medium (1,000 cps/µL), and low (100 cps/µL) concentrations (RUO)
Catalog No. NC3949383
Encompass_Preferred
Individual VanA,VanB,VanC1,VanC2/3 synthetic quantitative standards are designed to support precise assay calibration and validation in molecular diagnostics. Supplied at three concentrations—high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL)—these standards enable accurate assessment of assay performance across a dynamic range. They are ideal for sensitivity testing, method development, quality control, and proficiency testing, ensuring reproducible and reliable detection of these target sequences. Ready-to-use and stable, these standards streamline workflow integration while maintaining high confidence in molecular assay results. Suitable for research, clinical, and surveillance applications, they provide a trusted reference for laboratories aiming for accuracy and consistency in nucleic acid detection. (RUO)
Catalog No. NC3949383 Supplier GENEPATH DIAGNOSTICS INC Supplier No. 10890040050
Individual VanA,VanB,VanC1,VanC2/3 synthetic quantitative standards are designed to support precise assay calibration and validation in molecular diagnostics. Supplied at three concentrations—high (10,000 copies/µL), medium (1,000 copies/µL), and low (100 copies/µL)—these standards enable accurate assessment of assay performance across a dynamic range. They are ideal for sensitivity testing, method development, quality control, and proficiency testing, ensuring reproducible and reliable detection of these target sequences. Ready-to-use and stable, these standards streamline workflow integration while maintaining high confidence in molecular assay results. Suitable for research, clinical, and surveillance applications, they provide a trusted reference for laboratories aiming for accuracy and consistency in nucleic acid detection. (RUO)