Learn More
Ted Pella Inc Individual Well, 11.2mm I.D. x 22.3mm Deep, 420µm opening screen, all polypropylene
SDP
Catalog No. NC0664779
Encompass
Great for processing individual specimens or keeping specimens separate from others. Same wells as used in the Wellplate Inserts above but on an individual basis. Made entirely from polypropylene.
Catalog No. NC0664779 Supplier Ted Pella Inc Supplier No. 36173
Great for processing individual specimens or keeping specimens separate from others. Same wells as used in the Wellplate Inserts above but on an individual basis. Made entirely from polypropylene.