Learn More
MOLDED PRODUCTS INC MOLDED PRODUCTS INC Plastic Hemostat, Tube Occluding Forceps, Individually Packaged, X-Ray Detectable, Latex-Free
Catalog No. NC3973896
Encompass_Preferred
Medical tube occluding clamps for procedures. Also known as occluding forceps, they manipulate tubes without damage. Features a superior Maxi-Lock System.
  • Controls fluid flow in IV therapy.
  • Occludes dialysis tubing.
  • Clamps tubing for lab procedures.
  • Precision locking mechanism prevents slippage.
  • Color holds, no “chalk-out”.
  • X-ray detectable.
  • Comfortable oval finger holes for gloved hands.
  • Smooth jaws protect tubing.
  • Jaws remain aligned.
  • Safe, effective, and easy to use.
  • Latex-free.
Catalog No. NC3973896 Supplier MOLDED PRODUCTS INC Supplier No. MPC200Y1
Medical tube occluding clamps for procedures. Also known as occluding forceps, they manipulate tubes without damage. Features a superior Maxi-Lock System.
  • Controls fluid flow in IV therapy.
  • Occludes dialysis tubing.
  • Clamps tubing for lab procedures.
  • Precision locking mechanism prevents slippage.
  • Color holds, no “chalk-out”.
  • X-ray detectable.
  • Comfortable oval finger holes for gloved hands.
  • Smooth jaws protect tubing.
  • Jaws remain aligned.
  • Safe, effective, and easy to use.
  • Latex-free.