Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Individually Wrapped Hand Sanitizing Wipes Alcohol Formula
SDP

Catalog No. 19044053 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Packaging Quantity:
100 Wipes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-044-053 100 Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-053 Supplier PURELL™ Supplier No. 902210

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hand Sanitizing Wipes Alcohol Formula

  • Hand Sanitizing Wipes
  • Alcohol Formula
  • Fragrance Free
  • 100 Count
  • Individually Wrapped
TRUSTED_SUSTAINABILITY

Specifications

Product Type Interfold Wiper
Length (Metric) 7.81 cm
Width (Metric) 4.75 cm
For Use With (Application) Hand Sanitizing Wipe
Packaging Quantity 100 Wipes
Saturation Solution Ethyl Alcohol
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.