Learn More
Biotium Indo-1, AM Ester, 10x100 ug
Catalog No. 501964700
Encompass_Preferred
Indo-1, AM ester is membrane-permeant and thus can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate cell loading. Indo-1/AM itself does not bind Ca2 , but it is readily hydrolyzed to Indo-1 by endogenous esterases once it enters cells. Properties: Light yellow solid soluble in DMSO; Store at -20°C and protect from light, especially in solution; C47H51N3O22; MW: 1010; [112926-02-0]
Catalog No. 50-196-4700 Supplier Biotium Supplier No. 50043
Indo-1, AM ester is membrane-permeant and thus can be loaded into cells via incubation. Because of the relatively low water solubility of the AM ester, Pluronic F-127, a mild detergent, is often used as a dispersing agent to facilitate cell loading. Indo-1/AM itself does not bind Ca2 , but it is readily hydrolyzed to Indo-1 by endogenous esterases once it enters cells. Properties: Light yellow solid soluble in DMSO; Store at -20°C and protect from light, especially in solution; C47H51N3O22; MW: 1010; [112926-02-0]