Learn More
Biotium Indo-1, Pentapotassium Salt
Catalog No. 501964698
Encompass_Preferred
Membrane-impermeant calcium indicator that can be loaded into cells via microinjection or scrape loading. Properties, Kd: 250 nM; Ex/Em (no Ca2 ) = 349/482 nm; Ex/Em (high Ca2) = 331/398 nm; e(349 nm, no Ca2) = 34,000 M-1cm-1; Light yellow solid soluble in DMSO; Store at 4°C and protect from light, especially in solution; C32H26K5N3O12; MW: 840; [132319-56-3]
Catalog No. 50-196-4698 Supplier Biotium Supplier No. 50041
Membrane-impermeant calcium indicator that can be loaded into cells via microinjection or scrape loading. Properties, Kd: 250 nM; Ex/Em (no Ca2 ) = 349/482 nm; Ex/Em (high Ca2) = 331/398 nm; e(349 nm, no Ca2) = 34,000 M-1cm-1; Light yellow solid soluble in DMSO; Store at 4°C and protect from light, especially in solution; C32H26K5N3O12; MW: 840; [132319-56-3]