Learn More
AdipoGen Indo-1 pentapotassium salt
SDP
Catalog No. 502560669
Encompass_Preferred
Chemical. CAS 132319-56-3. Formula C32H26K5N3O12. MW 840.05. Synthetic. Fluorescent calcium chelator and indicator with an unusually large shift in fluorescence emission from 480 nm to 400 nm on binding of calcium. Useful for intracellular application.
Catalog No. 50-256-0669 Supplier AdipoGen Supplier No. CDXI0018M002
Chemical. CAS 132319-56-3. Formula C32H26K5N3O12. MW 840.05. Synthetic. Fluorescent calcium chelator and indicator with an unusually large shift in fluorescence emission from 480 nm to 400 nm on binding of calcium. Useful for intracellular application.