Learn More
AdipoGen Indocyanine Green
SDP
Catalog No. 502560673
Encompass_Preferred
Chemical. CAS 3599-32-4. Formula C43H47N2NaO6S2. MW 774.98. Indocyanine green ICG is a cyanine dye. It is a clinically approved near infrared NIR fluorescent dye and used in medical diagnostics, in vitro, in vivo and animal model study. NIR fluorescence is being utilized in a wide range of research fields. ICG is a vital fluorescent dye that binds to plasma proteins and lipoproteins. It displays excitation maxima ranging from 750 to 800 nm and an emission maximum of greater than or equal to 800 nm, which shifts from 810-820 nm in aqueous solution to 820-834 nm following intravenous injection. Formulations containing indocyanine green have been commonly used in clinical and surgical applications, including retinal angiography, intraocular surgery, and cardiac output and liver condition monitoring. ICG might be excited using 750-800 nm laser line or LED and displays excellent optical properties.
Catalog No. 50-256-0673 Supplier AdipoGen Supplier No. CDXI0013M500
Chemical. CAS 3599-32-4. Formula C43H47N2NaO6S2. MW 774.98. Indocyanine green ICG is a cyanine dye. It is a clinically approved near infrared NIR fluorescent dye and used in medical diagnostics, in vitro, in vivo and animal model study. NIR fluorescence is being utilized in a wide range of research fields. ICG is a vital fluorescent dye that binds to plasma proteins and lipoproteins. It displays excitation maxima ranging from 750 to 800 nm and an emission maximum of greater than or equal to 800 nm, which shifts from 810-820 nm in aqueous solution to 820-834 nm following intravenous injection. Formulations containing indocyanine green have been commonly used in clinical and surgical applications, including retinal angiography, intraocular surgery, and cardiac output and liver condition monitoring. ICG might be excited using 750-800 nm laser line or LED and displays excellent optical properties.