Learn More
Cayman Chemical Indocyanine Green, Purity: ≥95%, CAS: 3599-32-4, 100 mg
SDP
Catalog No. 502940465
Encompass_Preferred
A vital fluorescent dye; binds to plasma proteins and lipoproteins; ex/em = 750-800/≥800 nm, respectively, with emission maximum shifting from 810-820 nm in aqueous solution to 820-834 nm following intravenous injection
Catalog No. 50-294-0465 Supplier Cayman Chemical Supplier No. 27462100 MG
A vital fluorescent dye; binds to plasma proteins and lipoproteins; ex/em = 750-800/≥800 nm, respectively, with emission maximum shifting from 810-820 nm in aqueous solution to 820-834 nm following intravenous injection