Learn More
Cayman Chemical IndocynIn Green 250mg
SDP
Catalog No. 502938046
Encompass_Preferred
A vital fluorescent dye; binds to plasma proteins and lipoproteins; ex/em = 750-800/≥800 nm, respectively, with emission maximum shifting from 810-820 nm in aqueous solution to 820-834 nm following intravenous injection
Catalog No. 50-293-8046 Supplier Cayman Chemical Supplier No. 27462250 MG
A vital fluorescent dye; binds to plasma proteins and lipoproteins; ex/em = 750-800/≥800 nm, respectively, with emission maximum shifting from 810-820 nm in aqueous solution to 820-834 nm following intravenous injection