Learn More
AdipoGen (-)-Indolactam V,Chemical. CAS: 90365-57-4
SDP
Catalog No. 50464617
Encompass_Preferred
(-)-Indolactam V, AdipoGen Life Sciences. Chemical. CAS: 90365-57-4. Formula: C17H23N3O2. MW: 301.4. Tumor promoter. Protein kinase C (PKC) inducer. Mitogenesis activator. Differentiation inducer.
Catalog No. 50-464-617 Supplier AdipoGen Supplier No. AGCR10041C300
Add to Cart
Add to Cart
(-)-Indolactam V, AdipoGen Life Sciences. Chemical. CAS: 90365-57-4. Formula: C17H23N3O2. MW: 301.4. Tumor promoter. Protein kinase C (PKC) inducer. Mitogenesis activator. Differentiation inducer.