Learn More
Enzo Life Sciences (+)-Indolactam V (300µg). CAS: 90365-56-3
SDP
Supplier:  Enzo Life Sciences ALX420010C300
Encompass_Preferred
Used as negative control compound in studies with (-)-indolactam V (ALX-420-011). Purity: ≥95% (1H-NMR). Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-9096