Learn More
STEMCELL Technologies (-)-Indolactam V, Size: 1 mg
SDP
Supplier:  STEMCELL Technologies 72314
Encompass
(-)-Indolactam V is an indole alkaloid compound that activates protein kinase C (PKC). It binds to the α, β, γ, δ, ε, and η isozymes of PKC with Ki values of 11, 6, 19, 8, 22, and 16 nM respectively. (Kazanietz et al., Masuda et al.)DIFFERENTIATION· Promotes differentiation to human and mouse pancreatic precursors from pluripotent stem cell-derived definitive endoderm (Borowiak et al., Chen et al., Thatava et al.).
Catalog No. 50-197-5903