Learn More
Medchemexpress LLC Indole | 120-72-9 | 117.15 | 1 G
SDP
Catalog No. 5000447569
Encompass_Preferred
Indole is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. It regulates bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence, acting as an intercellular signal molecule.
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Widely distributed in the natural environment
  • Produced by various bacteria
Catalog No. 50-004-47569 Supplier Medchemexpress LLC Supplier No. HYW0011321G
Indole is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. It regulates bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence, acting as an intercellular signal molecule.
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Widely distributed in the natural environment
  • Produced by various bacteria