Learn More
Medchemexpress LLC Indole | 120-72-9 | 117.15 | 1 ML
SDP
Catalog No. 5000449556
Encompass_Preferred
Indole is an aromatic, heterocyclic, organic compound found extensively in the natural environment. Produced by various bacteria, it functions as an intercellular signal molecule, regulating bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence.
  • Aromatic, heterocyclic, organic compound
  • Widely distributed in the natural environment
  • Produced by various bacteria
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Inhibits cell proliferation
  • Moderates cell cycle progression
Catalog No. 50-004-49556 Supplier Medchemexpress LLC Supplier No. HYW00113210MM/1ML
Indole is an aromatic, heterocyclic, organic compound found extensively in the natural environment. Produced by various bacteria, it functions as an intercellular signal molecule, regulating bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence.
  • Aromatic, heterocyclic, organic compound
  • Widely distributed in the natural environment
  • Produced by various bacteria
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Inhibits cell proliferation
  • Moderates cell cycle progression