Learn More
Medchemexpress LLC Indole | 120-72-9 | 117.15 | 50 MG
SDP
Catalog No. 5000446913
Encompass_Preferred
Indole is an aromatic, heterocyclic, organic compound found widely in nature and produced by various bacteria. It acts as an intercellular signal molecule, regulating bacterial physiology including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence.
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Inhibits cell proliferation in certain cell lines
  • Increases protein expression of occludin and claudin-3
Catalog No. 50-004-46913 Supplier Medchemexpress LLC Supplier No. HYW00113250MG
Indole is an aromatic, heterocyclic, organic compound found widely in nature and produced by various bacteria. It acts as an intercellular signal molecule, regulating bacterial physiology including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence.
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Inhibits cell proliferation in certain cell lines
  • Increases protein expression of occludin and claudin-3