Learn More
Medchemexpress LLC Indole-13C | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW001132S1
Encompass_Preferred
Indole-13C is a 13C-labeled Indole, an endogenous metabolite. Indole is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. It regulates aspects of bacterial physiology, including spore formation, plasmid stability, resistance to drugs, biofilm formation, and virulence, acting as an intercellular signal molecule.
  • Regulates spore formation
  • Regulates plasmid stability
  • Regulates resistance to drugs
  • Regulates biofilm formation
  • Regulates virulence
  • Acts as an intercellular signal molecule
Catalog No. 50-004-46713