Learn More
Medchemexpress LLC Indole-13C | 85597-55-3 | 118.14 | 10 MG
SDP
Catalog No. 5000446622
Encompass_Preferred
Indole-13C is the 13C labeled Indole. Indole is an endogenous metabolite. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • 13C labeled indole
  • Endogenous metabolite
  • Suitable for research use
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-46622 Supplier Medchemexpress LLC Supplier No. HYW001132S110MG
Indole-13C is the 13C labeled Indole. Indole is an endogenous metabolite. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • 13C labeled indole
  • Endogenous metabolite
  • Suitable for research use
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS