Learn More
Medchemexpress LLC Indole-13C | 85597-55-3 | 118.14 | 100 MG
SDP
Catalog No. 5000449555
Encompass_Preferred
Indole-13C is the 13C labeled Indole. Indole is an endogenous metabolite.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-49555 Supplier Medchemexpress LLC Supplier No. HYW001132S1100MG
Indole-13C is the 13C labeled Indole. Indole is an endogenous metabolite.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.