Learn More
Medchemexpress LLC Indole-13C | 85597-55-3 | 118.14 | 5 MG
SDP
Catalog No. 5000447676
Encompass_Preferred
Indole-13C is the 13C labeled Indole, which is an endogenous metabolite. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes like Indole-13C are incorporated into drug molecules as tracers for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-47676 Supplier Medchemexpress LLC Supplier No. HYW001132S15MG
Indole-13C is the 13C labeled Indole, which is an endogenous metabolite. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes like Indole-13C are incorporated into drug molecules as tracers for quantitation during drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS