Learn More
Medchemexpress LLC Indole-2-carboxylic acid | 1477-50-5 | MFCD00005611 | 99.8% | 161.16 g/mol | C9H7NO2 | 10 MG
SDP
Catalog No. 5000297570
Encompass_Preferred
Indole-2-carboxylic acid analytical standard used as a reference material for chromatographic and analytical assays. Supplied as a high-purity solid for use in HPLC method development, calibration, and quality control.
  • Suitable as an analytical reference standard for HPLC and GC.
  • High purity confirmed by HPLC (99.78%).
  • Solid form and stable under recommended storage conditions.
  • Supplied in a brown glass bottle for preservation of sample integrity.
  • Useful for method development, calibration, and QC applications.
Catalog No. 50-002-97570 Supplier Medchemexpress LLC Supplier No. HYI0096R10MG
Indole-2-carboxylic acid analytical standard used as a reference material for chromatographic and analytical assays. Supplied as a high-purity solid for use in HPLC method development, calibration, and quality control.
  • Suitable as an analytical reference standard for HPLC and GC.
  • High purity confirmed by HPLC (99.78%).
  • Solid form and stable under recommended storage conditions.
  • Supplied in a brown glass bottle for preservation of sample integrity.
  • Useful for method development, calibration, and QC applications.