Learn More
Medchemexpress LLC Indole-2-carboxylic acid derivative (dimethylsulfamoyl piperazinyl, morpholinoethyl, naphthyloxypropyl) | 1668553-26-1 | MFCD28399118 | 99.9% | 850.04 g/mol | C46H55N7O7S | 1MG
SDP
Catalog No. 5000237743
Encompass_Preferred
A-1210477 is a potent, selective small-molecule inhibitor of the anti-apoptotic protein MCL-1. It binds MCL-1 with high affinity (reported Ki ≈ 0.45 nM) and induces apoptosis in MCL-1-dependent cancer cell lines. Supplied as a high-purity solid for preclinical and in vitro research, it is intended for mechanistic studies of apoptosis and evaluation of MCL-1-targeted therapeutic strategies.
  • Potent MCL-1 inhibition with Ki ≈ 0.45 nM.
  • Induces apoptosis in MCL-1-dependent cancer cells.
  • High chemical purity suitable for in vitro studies.
  • Solid form with recommended cold storage for long-term stability.
  • Useful for mechanistic studies and preclinical evaluation of MCL-1 targeting.
Catalog No. 50-002-37743 Supplier Medchemexpress LLC Supplier No. HY124681MG
A-1210477 is a potent, selective small-molecule inhibitor of the anti-apoptotic protein MCL-1. It binds MCL-1 with high affinity (reported Ki ≈ 0.45 nM) and induces apoptosis in MCL-1-dependent cancer cell lines. Supplied as a high-purity solid for preclinical and in vitro research, it is intended for mechanistic studies of apoptosis and evaluation of MCL-1-targeted therapeutic strategies.
  • Potent MCL-1 inhibition with Ki ≈ 0.45 nM.
  • Induces apoptosis in MCL-1-dependent cancer cells.
  • High chemical purity suitable for in vitro studies.
  • Solid form with recommended cold storage for long-term stability.
  • Useful for mechanistic studies and preclinical evaluation of MCL-1 targeting.