Learn More
Apexbio Technology LLC INDOLE-3-ACETAMIDE-200MG
SDP
Catalog No. 5000569963
Encompass_Preferred
Indole-3-acetamide (CAS No 879-37-8) is a chemical compound of interest in the field of plant physiology and biotechnology It is an important intermediate in the biosynthesis of indole-3-acetic acid a major plant hormone The compound influences plant growth and development by regulating pathways associated with cell division and elongation In research Indole-3-acetamide exhibits activity within the nanomolar to low micromolar range making it valuable in various studies related to plant biology It is commonly used in vitro to investigate its role in auxin production and signal transduction pathways Experimental concentrations typically depend on the specific research design and it is frequently utilized in cell models and drug screening assays The compound s ability to modulate plant hormone levels also makes it relevant in agricultural studies aimed at enhancing crop yield and stress resistance
Catalog No. 50-005-69963 Supplier Apexbio Technology LLC Supplier No. C7163200
Add to Cart
Add to Cart
Indole-3-acetamide (CAS No 879-37-8) is a chemical compound of interest in the field of plant physiology and biotechnology It is an important intermediate in the biosynthesis of indole-3-acetic acid a major plant hormone The compound influences plant growth and development by regulating pathways associated with cell division and elongation In research Indole-3-acetamide exhibits activity within the nanomolar to low micromolar range making it valuable in various studies related to plant biology It is commonly used in vitro to investigate its role in auxin production and signal transduction pathways Experimental concentrations typically depend on the specific research design and it is frequently utilized in cell models and drug screening assays The compound s ability to modulate plant hormone levels also makes it relevant in agricultural studies aimed at enhancing crop yield and stress resistance