Learn More
Cayman Chemical Indole-3-acetamide, Purity: ≥98%, CAS: 879-37-8, 50 g
SDP
Catalog No. 502950272
Encompass_Preferred
An endogenous metabolite of tryptophan in phytopathogenic bacteria; exogenous application increases the expression of ami1 in Arabidopsis at 20 µM; reduces relative primary root elongation in Arabidopsis mutants that have increased amidase activity and lower indole-3-acetamide levels, but not in wild-type Arabidopsis, at 1 and 10 µM; inhibits mouse and rat liver, as well as P. fluorescens, tryptophan 2,3-dioxygenase
Catalog No. 50-295-0272 Supplier Cayman Chemical Supplier No. 3437450 G
An endogenous metabolite of tryptophan in phytopathogenic bacteria; exogenous application increases the expression of ami1 in Arabidopsis at 20 µM; reduces relative primary root elongation in Arabidopsis mutants that have increased amidase activity and lower indole-3-acetamide levels, but not in wild-type Arabidopsis, at 1 and 10 µM; inhibits mouse and rat liver, as well as P. fluorescens, tryptophan 2,3-dioxygenase