Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetamide | 879-37-8 | MFCD00005641 | 100.0% | 174.20 g/mol | C10H10N2O | 1 ML
SDP

Catalog No. 5000299866
Encompass_Preferred

Indole-3-acetamide is an endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA), a principal plant auxin. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research use in studies of auxin biosynthesis, plant physiology, and biochemical assays. The solution format offers convenience and consistent concentration for small-scale experiments.

  • Ready-to-use 10 mM solution in DMSO.
  • High purity, approximately 99.95%.
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • Soluble in DMSO; limited solubility in water; sonication or warming may be required for aqueous preparations.
  • Solution storage: keep at -80°C for up to 6 months or at -20°C for short-term storage.

Catalog No. 50-002-99866 Supplier Medchemexpress LLC Supplier No. HYW01678410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide is an endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA), a principal plant auxin. Supplied as a ready-to-use 10 mM solution in DMSO (1 mL), it is intended for research use in studies of auxin biosynthesis, plant physiology, and biochemical assays. The solution format offers convenience and consistent concentration for small-scale experiments.

  • Ready-to-use 10 mM solution in DMSO.
  • High purity, approximately 99.95%.
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • Soluble in DMSO; limited solubility in water; sonication or warming may be required for aqueous preparations.
  • Solution storage: keep at -80°C for up to 6 months or at -20°C for short-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.