Learn More
Medchemexpress LLC Indole-3-acetamide | 879-37-8 | MFCD00005641 | 174.20 g/mol | C10H10N2O | 1 G
SDP
Catalog No. 5000299958
Encompass_Preferred
Indole-3-acetamide is an indole derivative and a biosynthetic intermediate in the formation of indole-3-acetic acid (IAA), a primary plant auxin. Supplied as a dry powder for research and analytical use, it is commonly used as a metabolite standard, for auxin pathway studies, and in analytical method development.
  • Biosynthetic intermediate of indole-3-acetic acid (IAA).
  • Suitable as a metabolite or analytical standard for biochemical and plant biology research.
  • Provided as a powder with defined storage recommendations for stability.
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • High solubility in DMSO; limited solubility in water-ultrasonic or warming may be required.
  • Manufacturer datasheet provides solubility, storage, and handling information.
Catalog No. 50-002-99958 Supplier Medchemexpress LLC Supplier No. HYW0167841G
Indole-3-acetamide is an indole derivative and a biosynthetic intermediate in the formation of indole-3-acetic acid (IAA), a primary plant auxin. Supplied as a dry powder for research and analytical use, it is commonly used as a metabolite standard, for auxin pathway studies, and in analytical method development.
  • Biosynthetic intermediate of indole-3-acetic acid (IAA).
  • Suitable as a metabolite or analytical standard for biochemical and plant biology research.
  • Provided as a powder with defined storage recommendations for stability.
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol.
  • High solubility in DMSO; limited solubility in water-ultrasonic or warming may be required.
  • Manufacturer datasheet provides solubility, storage, and handling information.