Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetamide | 879-37-8 | MFCD00005641 | 99.9% | 174.20 g/mol | C10H10N2O | 250 MG
SDP

Catalog No. 5000302381
Encompass_Preferred

Indole-3-acetamide is an endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied as a research-grade powder or solution, it is intended for analytical and biological studies of auxin biosynthesis and metabolic pathways. The material has documented solubility characteristics and recommended storage conditions for long-term stability.

  • Endogenous metabolite and IAA biosynthesis intermediate
  • High purity (manufacturer batches ~99.9%)
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol
  • Available as solid or pre-made solution for research use
  • Soluble in DMSO and water with documented concentrations
  • Stable when stored at recommended temperatures for powder and solutions
  • Suitable for metabolic pathway and analytical assays

Catalog No. 50-003-02381 Supplier Medchemexpress LLC Supplier No. 250MGHYW016784R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide is an endogenous metabolite and a biosynthetic intermediate of indole-3-acetic acid (IAA). Supplied as a research-grade powder or solution, it is intended for analytical and biological studies of auxin biosynthesis and metabolic pathways. The material has documented solubility characteristics and recommended storage conditions for long-term stability.

  • Endogenous metabolite and IAA biosynthesis intermediate
  • High purity (manufacturer batches ~99.9%)
  • Molecular formula C10H10N2O; molecular weight 174.20 g/mol
  • Available as solid or pre-made solution for research use
  • Soluble in DMSO and water with documented concentrations
  • Stable when stored at recommended temperatures for powder and solutions
  • Suitable for metabolic pathway and analytical assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.