Learn More
Medchemexpress LLC Indole-3-acetamide | 879-37-8 | MFCD00005641 | 99.9% | 174.20 g·mol⁻¹ | C10H10N2O | 50 MG
SDP
Catalog No. 5000301212
Encompass_Preferred
Indole-3-acetamide is an endogenous metabolite and an intermediate in the biosynthesis of indole-3-acetic acid. This product is supplied as a high-purity analytical standard intended for research and analytical applications such as metabolite identification, reference standards, and method development.
  • High purity suitable for analytical applications.
  • Molecular formula C10H10N2O and molecular weight 174.20 g·mol⁻¹.
  • Provided as a 50 mg packaged solid for small-scale analytical use.
  • Suitable for metabolite identification and reference standard workflows.
  • Includes batch purity documentation for traceability.
Catalog No. 50-003-01212 Supplier Medchemexpress LLC Supplier No. HYW016784R50MG
Indole-3-acetamide is an endogenous metabolite and an intermediate in the biosynthesis of indole-3-acetic acid. This product is supplied as a high-purity analytical standard intended for research and analytical applications such as metabolite identification, reference standards, and method development.
  • High purity suitable for analytical applications.
  • Molecular formula C10H10N2O and molecular weight 174.20 g·mol⁻¹.
  • Provided as a 50 mg packaged solid for small-scale analytical use.
  • Suitable for metabolite identification and reference standard workflows.
  • Includes batch purity documentation for traceability.