Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetamide-d5 | 1204700-53-7 | MFCD08705083 | 100.0% | 179.23 g·mol⁻¹ | C10H5D5N2O | 1 MG
SDP

Catalog No. 5000301224
Encompass_Preferred

Indole-3-acetamide-d5 is the deuterium-labeled (d5) analogue of indole-3-acetamide, provided as a high-purity stable isotope standard for analytical, metabolic, and tracer studies. Supplied as a white to off-white solid, it has defined storage recommendations to preserve isotopic and chemical integrity for long-term use.

  • Deuterium-labeled analogue (d5) for isotope tracing and quantitation.
  • High purity (99.99%) suitable for analytical standards.
  • Molecular formula C10H5D5N2O; molecular weight 179.23 g·mol⁻¹.
  • Off-white solid appearance for easy handling and storage.
  • Available in small quantities (mg scale) for reference standards.
  • Recommended storage: powder at -20°C for long term, 4°C for short term.

Catalog No. 50-003-01224 Supplier Medchemexpress LLC Supplier No. HYW016784S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide-d5 is the deuterium-labeled (d5) analogue of indole-3-acetamide, provided as a high-purity stable isotope standard for analytical, metabolic, and tracer studies. Supplied as a white to off-white solid, it has defined storage recommendations to preserve isotopic and chemical integrity for long-term use.

  • Deuterium-labeled analogue (d5) for isotope tracing and quantitation.
  • High purity (99.99%) suitable for analytical standards.
  • Molecular formula C10H5D5N2O; molecular weight 179.23 g·mol⁻¹.
  • Off-white solid appearance for easy handling and storage.
  • Available in small quantities (mg scale) for reference standards.
  • Recommended storage: powder at -20°C for long term, 4°C for short term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.