Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetamide-d5 | 1204700-53-7 | MFCD08705083 | 100.0% | 179.23 g/mol | C10H5D5N2O | 10 MG
SDP

Catalog No. 5000299864
Encompass_Preferred

Indole-3-acetamide-d5 is the deuterium-labeled form of indole-3-acetamide used as a stable isotope standard for research and analytical applications, especially in studies of auxin (indole-3-acetic acid) biosynthesis and quantitative mass spectrometry.

  • Used as a stable isotope internal standard for quantitative analysis.
  • Deuterium-labeled; molecular formula C10H5D5N2O, molecular weight 179.23 g/mol.
  • High purity (99.99%).
  • White to off-white solid appearance.
  • Powder stable at -20°C for up to 3 years under recommended storage.

Catalog No. 50-002-99864 Supplier Medchemexpress LLC Supplier No. HYW016784S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetamide-d5 is the deuterium-labeled form of indole-3-acetamide used as a stable isotope standard for research and analytical applications, especially in studies of auxin (indole-3-acetic acid) biosynthesis and quantitative mass spectrometry.

  • Used as a stable isotope internal standard for quantitative analysis.
  • Deuterium-labeled; molecular formula C10H5D5N2O, molecular weight 179.23 g/mol.
  • High purity (99.99%).
  • White to off-white solid appearance.
  • Powder stable at -20°C for up to 3 years under recommended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.