Learn More
Medchemexpress LLC Indole-3-acetic acid-2,2-d2 | 24420-86-8 | 99.1% | 177.20 | C10H7D2NO2 | 10 MG
SDP
Catalog No. 5000295454
Encompass_Preferred
3-Indoleacetic acid-2,2-d2 is a deuterium-labeled analogue of indole-3-acetic acid used as an analytical standard and research reagent to study auxin biology and to support quantitative mass spectrometry assays.
  • Deuterium-labeled analog for use as an internal standard in mass spectrometry.
  • High chemical and isotopic purity for reliable quantitation.
  • Suitable for plant hormone research and cell-culture experiments.
  • Provided as a stable solid for easy handling and storage.
  • Available in small quantities tailored to analytical workflows.
Catalog No. 50-002-95454 Supplier Medchemexpress LLC Supplier No. HY18569S210MG
3-Indoleacetic acid-2,2-d2 is a deuterium-labeled analogue of indole-3-acetic acid used as an analytical standard and research reagent to study auxin biology and to support quantitative mass spectrometry assays.
  • Deuterium-labeled analog for use as an internal standard in mass spectrometry.
  • High chemical and isotopic purity for reliable quantitation.
  • Suitable for plant hormone research and cell-culture experiments.
  • Provided as a stable solid for easy handling and storage.
  • Available in small quantities tailored to analytical workflows.