Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-3-acetic acid, 99.2%, MP Biomedicals™

Catalog No. icn10203705 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10203705 5 g
ICN10203725 25 g
ICN10203780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10203705 Supplier MP Biomedicals, Inc Supplier No. 0210203705
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005636
InChI Key SEOVTRFCIGRIMH-UHFFFAOYSA-N
Synonym indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a
PubChem CID 802
ChEBI CHEBI:16411
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)CC1=CNC2=CC=CC=C12

Specifications

Color White
Quantity 5 g
Formula Weight 175.19
Percent Purity 0.992
Physical Form Powder
Chemical Name or Material Indole-3-acetic acid, Molecular biology and Cell biology
EINECSNumber 201-748-2
storageNote1 Protect from light
Recommended Storage Store at -0°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.