Learn More
Cayman Chemical Indole-3-acetIc AcId-d5 10mg
SDP
Catalog No. 502917030
Encompass_Preferred
An internal standard for the quantification of indole-3-acetic acid by GC- or LC-MS
Catalog No. 50-291-7030 Supplier Cayman Chemical Supplier No. 3889410 MG
An internal standard for the quantification of indole-3-acetic acid by GC- or LC-MS