Learn More
Bioworld Indole-3-Acetic Acid (IAA), 5 g
Catalog No. 50255046
Encompass_Preferred
Indole-3-acetic acid (IAA)Synonym: HeteroauxinAuxin growth regulatorStimulates root growthPlant tissue culture testedSoluble in ethanol or 1M NaOH
Catalog No. 50-255-046 Supplier Bioworld Supplier No. 306310101
Indole-3-acetic acid (IAA)Synonym: HeteroauxinAuxin growth regulatorStimulates root growthPlant tissue culture testedSoluble in ethanol or 1M NaOH