Learn More
Bioworld Indole-3-Acetic Acid (IAA), 25 g
Catalog No. 50255048
Encompass_Preferred
Indole-3-acetic acid (IAA)Synonym: HeteroauxinAuxin growth regulatorStimulates root growthPlant tissue culture testedSoluble in ethanol or 1M NaOH
Catalog No. 50-255-048 Supplier Bioworld Supplier No. 306310103
Indole-3-acetic acid (IAA)Synonym: HeteroauxinAuxin growth regulatorStimulates root growthPlant tissue culture testedSoluble in ethanol or 1M NaOH