Learn More
Sigma Aldrich Fine Chemicals Biosciences Indole-3-acetic acid sodium salt, 6505-45-9, MFCD09039281, 2 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I51482G
Encompass
Empirical Formula (Hill Notation): C10H8NNaO2, Molecular Weight: 197.17, plant cell culture tested, BioReagent, ≥98%, Synonym: Heteroauxin, IAA. Suitable for analytical standard, and elucidation of molecular pathways that mediate plant growth.
Catalog No. NC1137554