Learn More
Cayman Chemical Indole-3-acetIc AcIdodIum 1g
SDP
Catalog No. 502965623
Encompass_Preferred
An auxin plant growth regulator; reduces plant height and shoot growth in tomato plants at 10 and 50 µM; root saturation with indole-3-acetic acid-loaded nanoparticles increases rooting efficiency in P. elaegrifolia
Catalog No. 50-296-5623 Supplier Cayman Chemical Supplier No. 169541 G
An auxin plant growth regulator; reduces plant height and shoot growth in tomato plants at 10 and 50 µM; root saturation with indole-3-acetic acid-loaded nanoparticles increases rooting efficiency in P. elaegrifolia