Learn More
Cayman Chemical Indole-3-acetic Acid (sodium salt), Purity: ≥98%, CAS: 6505-45-9, 5 g
SDP
Catalog No. 502966089
Encompass_Preferred
An auxin plant growth regulator; reduces plant height and shoot growth in tomato plants at 10 and 50 µM; root saturation with indole-3-acetic acid-loaded nanoparticles increases rooting efficiency in P. elaegrifolia
Catalog No. 50-296-6089 Supplier Cayman Chemical Supplier No. 169545 G
An auxin plant growth regulator; reduces plant height and shoot growth in tomato plants at 10 and 50 µM; root saturation with indole-3-acetic acid-loaded nanoparticles increases rooting efficiency in P. elaegrifolia