Learn More
Cayman Chemical Indole-3-acetonitrile, Purity: ≥98%, CAS: 771-51-7, 25 g
SDP
Catalog No. 502917935
Encompass_Preferred
An indole alkaloid with antibacterial activity; reduces biofilm formation by E. coli and P. aeruginosa at 100 µg/ml; levels are increased in P. brassicae-infected cabbage roots
Catalog No. 50-291-7935 Supplier Cayman Chemical Supplier No. 3875125 G
An indole alkaloid with antibacterial activity; reduces biofilm formation by E. coli and P. aeruginosa at 100 µg/ml; levels are increased in P. brassicae-infected cabbage roots